missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ACE-2 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9332T-100UG
This item is not returnable.
View return policy
Description
ACE-2 Monoclonal specifically detects ACE-2 in Human samples. It is validated for Flow Cytometry.
Specifications
| ACE-2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
| Mouse myeloma cell line NS0-derived recombinant human ACE-2, Gln18-Ser740, Accession # Q9BYF1 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 535919 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9BYF1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 59272 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction