missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ACCSL (aa 87-175) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (46%), Rat (46%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139872 (PA5-139872. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ACCSL gene ontology annotations related to this gene include biosynthetic process.
Specifications
Specifications
| Accession Number | Q4AC99 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 390110 |
| Name | Human ACCSL (aa 87-175) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1-aminocyclopropane-1-carboxylate synthase (inactive)-like; 1-aminocyclopropane-1-carboxylate synthase homolog (Arabidopsis)(non-functional)-like; 1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like; 1-aminocyclopropane-1-carboxylate synthase-like protein 2; ACC synthase-like protein 2; ACCSL; Probable inactive 1-aminocyclopropane-1-carboxylate synthase-like protein 2 |
| Common Name | ACCSL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction