missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ABRA1 Control Fragment Recombinant Protein

Product Code. 30202045
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202045

Brand: Invitrogen™ RP102777

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (51%), Rat (51%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58042 (PA5-58042. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CCDC98, also known as Abraxas 1, was identified through protein binding studies using the breast and ovarian predisposition protein BRCA1 as the binding target. CCDC98 recruits RAP80, a ubiquitin-binding protein, to BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both CCDC98 and RAP80 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either CCDC98 or RAP80 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that CCDC98 and RAP80 control only part of the DNA damage response role of BRCA1. At least two isoforms of CCDC98 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6UWZ7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84142
Name Human ABRA1 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3830405G04Rik; 5630400M01Rik; ABRA1; abraxas protein; Abraxas1; AI506069; AL024423; AV118690; BRCA1-A complex subunit Abraxas; BRCA1-A complex subunit Abraxas 1; Ccdc98; coiled-coil domain containing 98; coiled-coil domain-containing protein 98; FAM175A; family with sequence similarity 175 member A; family with sequence similarity 175, member A; FLJ11520; FLJ12642; FLJ13614; Protein FAM175A; RGD1305287; UNQ496/PRO1013
Common Name ABRA1
Gene Symbol Abraxas1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CNYNHHLDVVDNLTLMVEHTDIPEASPASTPQIIKHKTLDLDDRWQFKRSRLLDTQDKRSKADTGSSNQDKASKMSSPETDEEIEKMKGFGEYSRSPT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.