missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ABCE1 (aa 178-265) Control Fragment Recombinant Protein

Product Code. 30210805
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210805

Brand: Invitrogen™ RP104669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111079 (PA5-111079. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the OABP subfamily. Alternatively referred to as the RNase L inhibitor, this protein functions to block the activity of ribonuclease L. Activation of ribonuclease L leads to inhibition of protein synthesis in the 2-5A/RNase L system, the central pathway for viral interferon action. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P61221
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6059
Name Human ABCE1 (aa 178-265) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2'-5'-oligoadenylate-binding protein; ABC 38; ABC38; ABCE 1; ABCE1; ATP binding cassette subfamily E member 1; ATP-binding cassette sub-family E member 1; ATP-binding cassette, subfamily E (OABP), member 1; ATP-binding cassette, sub-family E (OABP), member 1; ATP-binding cassette, subfamily E, member 1; C79080; HP68; HuHP68; OABP; OK/SW-cl0.40; Ribonuclease 4 inhibitor; ribonuclease L (2', 5'-oligoisoadenylate synthetase-dependent) inhibitor; ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent) inhibitor; RLI; RLI1; RNase 4 inhibitor; RNase L inhibitor; RNase L inhibitor 1; RNASEL1; Rnaseli; RNS41; RNS4I; RNS4l (Eye)
Common Name ABCE1
Gene Symbol ABCE1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KAAKGTVGSILDRKDETKTQAIVCQQLDLTHLKERNVEDLSGGELQRFACAVVCIQKADIFMFDEPSSYLDVKQRLKAAITIRSLINP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.