missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ABCA7 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
ABCA7 Synthetic Peptide, PEP-1438, from Invitrogen.
ATP-binding cassette (ABC) transporters are an evolutionarily conserved family that use ATP hydrolysis to catalyze the transport of various molecules across cell membranes. They are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). ABCA7, a member of the ABC1 subfamily, is a full-size ABC transporter consisting of two sets of the multiple membrane-spanning domains plus the Walker motifs for the ATP interaction. ABCA7 shows the highest homology to ABCA1, an essential molecule for cholesterol homeostasis. The high expression levels of ABCA7 in peripheral leukocytes, thymus, spleen and bone marrow suggests a role in the immune system lipid homeostasis.
Specifications
Specifications
| Gene ID (Entrez) | 10347 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q8IZY2 |
| Common Name | Human ABCA7 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction