missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AASDH (aa 440-533) Control Fragment Recombinant Protein

Product Code. 30205065
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205065

Brand: Invitrogen™ RP96699

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57674 (PA5-57674. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

AASDH encodes a member of the non-ribosome peptide syntesase (NRPS) enzyme family. The encoded protein contains an AMP-binding domain, PP-binding (phosphopantetheine, or pantetheine 4'phosphate-binding) domain and the Pyrrolo-quinoline quinon (PQQ) binding domain. The protein is expressed in several adult tissues. Covalently binds beta-alanine in an ATP-dependent manner to form a thioester bond with its phosphopantetheine group and transfers it to an, as yet, unknown acceptor. May be required for a post-translational protein modification or for post-transcriptional modification of an RNA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q4L235
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 132949
Name Human AASDH (aa 440-533) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2-aminoadipic 6-semialdehyde dehydrogenase; 2-aminoadipic 6-semialdehyde dehydrogenase antibody; A230062G08Rik; A830035E16; Aasdh; AASDH antibody; ACSF4; ACSF4 antibody; acyl-CoA synthetase family member 4; acyl-CoA synthetase family member 4 antibody; aminoadipate-semialdehyde dehydrogenase; aminoadipate-semialdehyde dehydrogenase antibody; Beta-alanine-activating enzyme; HSPC318; LYS2; LYS2 antibody; non-ribosomal peptide synthetase 1098; non-ribosomal peptide synthetase 1098 antibody; non-ribosomal peptide synthetase 998; non-ribosomal peptide synthetase 998 antibody; NRPS1098; NRPS1098 antibody; NRPS998; NRPS998 antibody; Protein LYS2 homolog; Protein NRPS998; putative aminoadipate-semialdehyde dehydrogenase; RGD1311135; U26
Common Name AASDH
Gene Symbol AASDH
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LGRKDSQIKRHGKRLNIELVQQVAEELQQVESCAVTWYNQEKLILFMVSKDASVKEYIFKELQKYLPSHAVPDELVLIDSLPFTSHGKIDVSEL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.