missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human A-Raf Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6607-SP
This item is not returnable.
View return policy
Description
A-RAF Monoclonal specifically detects A-RAF in Human samples. It is validated for Western Blot.
Specifications
| A-RAF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P10398 | |
| ARAF | |
| E. coli-derived recombinant human A-Raf Asn150-Lys314 Accession # P10398 | |
| 25 μg | |
| Primary | |
| Detects human A-Raf in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) B-Raf, rhRaf-1 (aa 1-648), rhRaf-1 (aa 189-353), or rhKSR1 is observed. In Western blots, no cross-reactivity with recombinant human (rh) B-Raf or rhKSR1 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 684128 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARaf, ARAF1A-RAF, EC 2.7.11, EC 2.7.11.1, Oncogene ARAF1, PKS, PKS2, PKS2A-Raf proto-oncogene serine/threonine-protein kinase, Proto-oncogene A-Raf, Proto-oncogene A-Raf-1, Proto-oncogene Pks, RAFA1, Ras-binding protein DA-Raf, serine/threonine-protein kinase A-Raf, v-raf murine sarcoma 3611 viral oncogene homolog, v-raf murine sarcoma 3611 viral oncogene homolog 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 369 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction