missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 5T4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4975
This item is not returnable.
View return policy
Description
5T4 Monoclonal specifically detects 5T4 in Human samples. It is validated for Immunohistochemistry.
Specifications
| 5T4 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q13641 | |
| TPBG | |
| Mouse myeloma cell line NS0-derived recombinant human 5T4 Ser31-Ser355 Accession # Q13641 | |
| 100 μg | |
| Primary | |
| Detects human 5T4 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse 5T4 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 524731 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, M6P1, TPBG, trophoblast glycoprotein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7162 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction