missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 5-HT4 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB6545T-100UG
This item is not returnable.
View return policy
Description
5-HT4 Monoclonal antibody specifically detects 5-HT4 in Human samples. It is validated for Flow Cytometry
Specifications
| 5-HT4 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3360 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 631769 | |
| Flow Cytometry | |
| 5HT4, 5-HT-4, 5-HT4R, 5-hydroxytryptamine (serotonin) receptor 4,5-hydroxytryptamine receptor 4,5-HT45-hydroxytryptamine4 receptor, cardiac 5-HT4 receptor, HTR4, Serotonin receptor 4 | |
| NS0 mouse myeloma cell line transfected with human 5-HT4 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction