missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 5-HT2C Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB5764N-100UG
This item is not returnable.
View return policy
Description
5-HT2C Monoclonal specifically detects 5-HT2C in Human samples. It is validated for Flow Cytometry.
Specifications
| 5-HT2C | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| 5-HT1C, 5-HT-1C, 5HT2C, 5-HT-2C, 5-HT2C5-hydroxytryptamine receptor 1C, 5-HTR2C, 5-hydroxytryptamine (serotonin) receptor 2C, 5-hydroxytryptamine receptor 2C, HTR1C, HTR2C, serotonin 5-HT-2C receptor, Serotonin receptor 2C | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human 5HT2C in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 496214 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| HTR2C | |
| NS0 mouse myeloma cell line transfected with human 5HT2C | |
| RUO | |
| 3358 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction