missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 5-HT2B Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£181.00 - £560.00
Specifications
| Antigen | 5-HT2B |
|---|---|
| Clone | 2880B |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702927
![]() |
R&D Systems
MAB11297-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773307
![]() |
R&D Systems
MAB11297-100 |
100 μg |
£560.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-HT2B Monoclonal antibody specifically detects 5-HT2B in Human samples. It is validated for Flow Cytometry, ImmunohistochemistrySpecifications
| 5-HT2B | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 5-HT(2B), 5HT2B, 5-HT-2B, 5-HT2B5-HT 2B receptor, 5-hydroxytryptamine (serotonin) receptor 2B, 5-hydroxytryptamine 2B receptor, 5-hydroxytryptamine receptor 2B, HTR2B, Serotonin receptor 2B | |
| Synthetic peptide containing human 5- HTR2B, Accession # P41595 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2880B | |
| Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P41595 | |
| 3357 | |
| IgG | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title