missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 4-1BB Ligand/TNFSF9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £397.00
Specifications
| Antigen | 4-1BB Ligand/TNFSF9 |
|---|---|
| Clone | 282220 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18781299
![]() |
R&D Systems
MAB2295-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18181571
![]() |
R&D Systems
MAB2295 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Especificaciones
| 4-1BB Ligand/TNFSF9 | |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P41273.1 | |
| 8744 | |
| E. coli-derived recombinant human 4-1BB Ligand/TNFSF9 Arg71-Glu254 Accession # P41273.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human 4-1BB Ligand/TNFSF9 Ligand in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) 4-1BB Ligand, recobinant human (rh) APRIL, rhBAFF, rmEDA-A2, rhFas Ligand, rhGITR Ligand, rhLIGHT, rhLymphotoxin alpha 1/beta 2, rhLymphotoxin alpha 2/beta 1, rhOX40 Ligand, rhTNF-alpha , rhTRAIL, rhTRANCE, or rhTWEAK is observed. |
| 282220 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 4-1BB ligand, 41BB Ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, TNFSF9, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
| TNFSF9 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto