missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 2B4/CD244/SLAMF4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1039-SP
This item is not returnable.
View return policy
Description
2B4/CD244/SLAMF4 Polyclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Agonist Activity, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| 2B4/CD244/SLAMF4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD244, CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAIL, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4, SLAMF4NK cell activation inducing ligand NAIL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects human 2B4/CD244/SLAMF4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant mouse 2B4, recombinant human (rh) SLAM, rhCD48, and rhCD84 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Agonist Activity, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| NP_057466 | |
| CD244 | |
| Mouse myeloma cell line NS0-derived recombinant human 2B4/CD244/SLAMF4 Cys22-Arg221 Accession # NP_057466 | |
| 25 μg | |
| Agonist Activity | |
| 51744 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction