missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human 20S Proteasome Subunit alpha-5 (aa 120-133) Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
20S Proteasome Subunit alpha5 (residues 120-133) Synthetic Peptide, PEP-210, from Invitrogen.
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit.
Specifications
Specifications
| Gene ID (Entrez) | 5686 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P28066 |
| Common Name | Human 20 S Proteasome Subunit alpha-5 (aa 120-133) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction