missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HuD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579199
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue, U87-MG whole cell. IHC: mouse brain tissue, rat brain tissue, human meningeoma tissue.
HuD, otherwise known as ELAV-like protein 4 or PNEM, is a protein that in humans is encoded by the ELAVL4 gene. This human gene is located at 1p34 by in situ hybridization. The HuD/ELAVL4 protein is an RNA-binding protein. ELAVL4 has specificity for 3-prime uridylate-rich untranslated regions of growth factor mRNAs. HuD is expressed only in neurons and it binds to AU-rich element-containing mRNAs. As a result of this interaction the, half-life of the transcript is increased. Additionally, HuD is important in neurons during brain development and plasticity.
Specifications
| HuD | |
| Polyclonal | |
| Unconjugated | |
| ELAVL4 | |
| Elav; ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like neuron-specific RNA binding protein 4; ELAVL4; ELAV-like protein 4; Hu antigen D; HU-antigen D; Hud; Paraneoplastic encephalomyelitis antigen HuD; PNEM; RGD1561943; r-HuD; RNA-binding protein HUD3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15572, 1996, 432358 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| O09032, P26378, Q61701 | |
| ELAVL4 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human ELAVL4 (8-45aa MEPQVSNGPTSNTSNGPSSNNRNCPSPMQTGATTDDSK). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction