missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUCE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87610
This item is not returnable.
View return policy
Description
HUCE1 Polyclonal specifically detects HUCE1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| HUCE1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23378 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry-Paraffin | |
| Cerebral protein 1, KIAA0409EC 2.1.1.-, NML, nucleomethylin, ribosomal RNA processing 8, methyltransferase, homolog (yeast), ribosomal RNA-processing protein 8, RRP8 methyltransferase homolog | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human HUCE1 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction