missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HuC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55047-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody has no cross reaction to other ELAVL members.
HUC, belongs to the RRM elav family. It binds to AU-rich sequences (AREs) of target mRNAs, including VEGF mRNA. ELAVL3 may also bind poly-A tracts via RRM 3. It is involved in neuronal differentiation and maintece.Specifications
| HuC | |
| Polyclonal | |
| Unconjugated | |
| ELAVL3 | |
| ELAV like protein 3, ELAVL3, Hu antigen C, HUC, HUCL, PLE21 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15571, 1995, 282824 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14576, Q60900 | |
| ELAVL3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction