missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HuC Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | HuC |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18352456
|
Novus Biologicals
NBP3-02943-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18352884
|
Novus Biologicals
NBP3-02943-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HuC Polyclonal specifically detects HuC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HuC | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp547J036, ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C), ELAV-like protein 3, Hu antigen C, Hu-antigen C, HuC, HUCL, HUCParaneoplastic cerebellar degeneration-associated antigen, MGC20653, PLE21Paraneoplastic limbic encephalitis antigen 21 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of humanHuC (NP_001411.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 1995 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title