Learn More
Herka™ Hubbard™ Borosilicate Glass Pycnometer
Product Code. 11900657
Change view
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
11900657 30 mL Each
1 options
Product Code. 11900657 Supplier Herka™ Supplier No. A1858300
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for determining and comparing density or specific gravity values. Herka™ Hubbard™ Borosilicate Glass Pycnometer is suitable for use with liquids or solids.
  • For use with liquids or solids
  • Borosilicate glass construction
Specifications
Capacity (Metric) 30 mL
Material Borosilicate Glass
For Use With (Application) Measuring density and specific gravity