missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HU 308

Product Code. 18065212
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18065212 50 mg 50mg
18023992 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065212 Supplier Tocris Bioscience™ Supplier No. 3088/50
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective CB2 agonist

Potent and selective CB2 receptor agonist (Ki values are 22.7 nM and >10 μM for CB2 and CB1 receptors respectively, EC50 = 5.57 nM). Displays antiallodynic activity in the rat hindpaw incision model of postoperative pain. Also neuroprotective and improves motor performance in a mouse model of Huntingtons Disease.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HU 308
CAS 256934-39-1
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C27H42O3
Formula Weight 414.62
Molecular Weight (g/mol) 414.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.