missing translation for 'onlineSavingsMsg'
Learn More

HTS01037, MedChemExpress

Product Code. 16453398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453398 10 mg 10mg
16449926 50 mg 50mg
16434636 100 mg 100mg
16416127 5 mg 5mg
16412756 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16453398 Supplier Medchem Express Supplier No. HY101503

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HTS01037 is an inhibitor of fatty acid binding; and a competitive antagonist of protein-protein interactions mediated by AFABP/aP2 with a Ki of 0.67 μM.

HTS01037 is an inhibitor of fatty acid binding

Specifications

Chemical Name or Material HTS01037
CAS 682741-29-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H11NO5S2
Quantity 10 mg
Solubility Information DMSO : ≥ 150 mg/mL (444.62 mM)
SMILES O=C(C1=C(NC(/C=C/C(O)=O)=O)C=C(C2=CC=CS2)S1)OC
Molecular Weight (g/mol) 337.37
Formula Weight 337.37
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.