missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTRA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560992
This item is not returnable.
View return policy
Description
Immunogen sequence: FAIPSDRVRQ FLAEYHEHQM KGKAFSNKKY LGLQMLSLTV PLSEELKMHY PDFPDVSSG Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 71%.
Serine protease.
Specifications
| HTRA4 | |
| Polyclonal | |
| Unconjugated | |
| HTRA4 | |
| B430206E18Rik; High-temperature requirement factor A4; HtrA serine peptidase 4; HTRA4; probable serine protease HTRA4; RGD1306242; serine protease HTR4; Serine protease HTRA4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 203100 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P83105 | |
| HTRA4 | |
| Recombinant protein corresponding to Human HTRA4. Recombinant protein control fragment (Product #RP-93896). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction