missing translation for 'onlineSavingsMsg'
Learn More

HTRA2/Omi Antibody, DyLight 405, Novus Biologicals™

Product Code. 18363342 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363342 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363342 Supplier Novus Biologicals Supplier No. NBP299409V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HTRA2/Omi Polyclonal specifically detects HTRA2/Omi in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTRA2/Omi
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant HTRA2/Omi (O43464-1; Ala134-Glu458).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Mitochondrial Mediated Pathway
Primary or Secondary Primary
Gene ID (Entrez) 27429
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.