missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HTRA2/Omi Monoclonal antibody specifically detects HTRA2/Omi in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Formulation | 50mM Sodium Borate |
| Regulatory Status | RUO |
| Research Discipline | Apoptosis, Cancer, Mitochondrial Mediated Pathway |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?