missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTRA2/Omi Antibody (196C429), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-56558
This item is not returnable.
View return policy
Description
HTRA2/Omi Monoclonal specifically detects HTRA2/Omi in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HTRA2/Omi | |
| Monoclonal | |
| Unconjugated | |
| NP_713130.1 | |
| HTRA2 | |
| This monoclonal antibody was developed by immunizing mice with a synthetic peptide containing amino acids 335-350 of human HtrA2. | |
| 0.1 mg | |
| Apoptosis, Cancer, Mitochondrial Mediated Pathway | |
| 27429 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 196C429 | |
| Western Blot 2-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (Lee et al (2003)) | |
| High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody does not cross-react with mouse HtrA2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction