missing translation for 'onlineSavingsMsg'
Learn More

HTRA1/PRSS11 Antibody (275603) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30734341 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734341 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734341 Supplier Novus Biologicals Supplier No. FAB29161MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HTRA1/PRSS11 Monoclonal antibody specifically detects HTRA1/PRSS11 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTRA1/PRSS11
Applications Immunohistochemistry
Classification Monoclonal
Clone 275603
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias ARMD7, EC 3.4.21, EC 3.4.21.108, High-temperature requirement A serine peptidase 1, HtrA serine peptidase 1, L56, ORF480, Serine protease 11, serine, 11 (IGF binding)
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human HTRA1/PRSS11, Gln23-Pro480, Accession # Q92743
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5654
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.