missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102047
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HTR5B.
SR-5B, also designated 5-hydroxytryptamine (serotonin) receptor 5B (5-HT5B) and G protein-coupled receptor 134 (GPR134), is a receptor for the monoamine ligand serotonin (5-hydroxytryptamine, 5-HT). Serotonin is a neurotransmitter derived from serotonergic neurons in the central nervous system and enterochromaffin cells in the gastrointestinal tract. Serotonin actions are mediated by receptors that influence the biochemistry of depression, anxiety, sexuality and appetite. Rat SR-5B is present in serotonergic neurons in dorsal raphe (DR) and central superior nucleus (CS, median raphe nucleus). DR cell bodies showing SR-5B mRNA expression are abundant in the medial portions of the nucleus. CS coexpression of SR-5B receptor mRNA with serotonin transporter mRNA is high in the intermediate portions of the nucleus.
Specifications
| HTR5B | |
| Polyclonal | |
| Unconjugated | |
| Htr5b | |
| 5ht5b; 5-HT5B; 5-HT-5B; 5-hydroxytryptamine (serotonin) receptor 5B; 5-hydroxytryptamine receptor 5B; Htr5b; MR22; protein-tyrosine-phosphatase (EC 3.1.3.48) Ly-5 precursor (B-cell variant); REC17; Serotonin receptor 5B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15564, 79247 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31387, P35365 | |
| Htr5b | |
| A synthesized peptide derived from rat HTR5B(Accession P35365), corresponding to amino acid residues V279-M299. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction