missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR3C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599881
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HTR3C.
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit C of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. Genes encoding subunits C, D and E form a cluster on chromosome 3.
Specifications
| HTR3C | |
| Polyclonal | |
| Unconjugated | |
| HTR3C | |
| 5-HT3C; 5-HT3-C; 5-hydroxytryptamine (serotonin) receptor 3, family member C; 5-hydroxytryptamine (serotonin) receptor 3C, ionotropic; 5-hydroxytryptamine receptor 3 subunit C; 5-hydroxytryptamine receptor 3C; HTR3C; Serotonin receptor 3C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170572 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WXA8 | |
| HTR3C | |
| A synthesized peptide derived from human HTR3C(Accession Q8WXA8), corresponding to amino acid residues Q366-V416. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction