missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR2A (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577748
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
In platelets and gut, serotonin plays a major role in cardiovascular function and motility of the gastrointestinal tract, respectively. Serotonin mediates its effects through several of G protein coupled receptors, designated 5-HT receptors or alternatively SR receptors. The SR-2 receptors are comprised of three subtypes, SR-2A, SR-2B and SR-2C, which activate phospholipase C and release intracellular stores of calcium in response to serotonin. SR-2A has a specific role in tracheal smooth muscle contraction, bronchoconstriction and mediating aldosterone production, and it is also thought to play a role in several psychiatric disorders, including depression and schizophrenia. SR-2B is expressed in embryonic and adult cardiovascular tissues, gut and brain and plays an important role in the pathology of cardiac disorders. SR-2C is thought to mediate the effects of atypical antipsychotic drugs.
Specifications
| HTR2A (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| HTR2A | |
| 5-HT-2, 5-HT2A, Serotonin receptor 2A, Htr2a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15558, 29595, 3356 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry, Western Blot | |
| 0.9 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P14842, P28223, P35363 | |
| Htr2a | |
| Peptide (C)SEASNWTIDAENR, corresponding to amino acid residues 40-52 of rat 5-Hydroxytryptamine Receptor 2A | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction