Learn More
Invitrogen™ HTR1E Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532863
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Monkey, Bat, Elephant, Horse, Guinea pig (100%); Orangutan, Gibbon, Bovine (95%); Panda (89%); Opossum (84%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
G-protein coupled receptor for 5-hydroxytryptamine (serotonin). Also functions as a receptor for various alkaloids and psychoactive substances. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors, such as adenylate cyclase. Signaling inhibits adenylate cyclase activity.
Specifications
| HTR1E | |
| Polyclonal | |
| Unconjugated | |
| HTR1E | |
| 5-HT1E; 5-HT-1E; 5HT1E Receptor; 5-HT1e receptor; 5-hydroxytryptamine (serotonin) receptor 1E, G protein-coupled; 5-hydroxytryptamine receptor 1E; HT1e receptor; HTR1E; S31; serotonin receptor 1E | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100065574, 100379275, 3354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P28566, Q6VB83 | |
| HTR1E | |
| Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human 5HT1E Receptor. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Horse, Guinea Pig, Bat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.