missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR1D Polyclonal Antibody

HTR1D Polyclonal Antibody
Brand: Invitrogen PA533294
This item is not returnable.
View return policy
Description
HTR1D Polyclonal Antibody for IHC (P)
Specifications
| HTR1D | |
| Polyclonal | |
| Unconjugated | |
| HTR1D | |
| 5 - Hydroxytryptamine (serotonin) receptor 1D; 5HT1D; 5-HT1D; 5-HT-1D; 5HT1D Receptor; 5-HT1d receptor; 5-HT-1D-alpha; 5-HT1DR; 5-hydroxytryptamine (serotonin) receptor 1D; 5-hydroxytryptamine (serotonin) receptor 1D beta; 5-hydroxytryptamine (serotonin) receptor 1D, G protein-coupled; 5-hydroxytryptamine receptor 1D; AI853647; Gpcr14; G-protein coupled receptor 14; HT1d receptor; HT1DA; Htr1d; HTR1DA; Htr1db; HTRL; RDC4; RP23-32L5.3; serotonin 1D alpha receptor; Serotonin receptor 1D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3352 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P28221 | |
| HTR1D | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human 5HT1D Receptor. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction