Learn More
HTR1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553716
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VEKTGADTRH GASPAPQPKK SVNGESGSRN WRLGVESKAG GALCANGAVR QGDDGAALEV IEVHRVGNSK EHLPLPSEAG PTPCAPASFE RKNERNAEAK RKMALA Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 72%.
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.Specifications
HTR1A | |
Polyclonal | |
Unconjugated | |
HTR1A | |
5HT1A; 5-HT1A; 5-HT-1A; 5-HT1a receptor; 5-hydroxytryptamine (serotonin) receptor 1A; 5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled; 5-hydroxytryptamine receptor 1A; ADRB2RL1; ADRBRL1; G-21; Gpcr18; G-protein coupled receptor 18; guanine nucleotide-binding regulatory protein-coupled receptor; HTR1A; PFMCD; RAT5HT1A; Serotonin receptor 1A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3350 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P08908 | |
HTR1A | |
Recombinant protein corresponding to Human HTR1A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only