missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals HTLV Blot 2.4 (RUO)
Shop All MP Biomedicals Products
Click to view available options
Quantity:
18 Tests/Kit
36 Tests/Kit
Unit Size:
1 set
Description
- The nitrocellulose strips are precoated with HTLV viral proteins derived from native inactivated viral particles and genetically engineered proteins.
- Individual nitrocellulose strips are incubated with diluted serum or plasma specimens and controls.
- Specific antibodies to HTLV-I/II, if present in the specimen will bind to the HTLV-I/II proteins on the strips.
- The strips are washed to remove unbound materials while antibodies that bind specifically to the HTLV proteins can be visualized using a series of reactions with goat anti-human IgG conjugated with alkaline phosphatase and the substrate, BCIP/NBT.
- Each strip also includes an internal sample addition control to minimize the risk of false negatives due to operational errors
- The possible serological profiles defined by the test include the following: HTLV Seropositive, HTLV-I Seropositive, HTLV-II Seropositive, Seronegative or Indeterminate.
- It is intended for use as a more specific supplemental test on human serum or plasma specimens found repeatedly reactive using screening procedures like ELISA.
- Research use only.
- For professional use only.
- Human serum and plasma.
Specifications
Specifications
| Product Type | Western Blot Kit |
| Detection Method | Colorimetric |
| For Use With (Application) | Immunoblot (Western) |
| Membrane Compatibility | Nitrocellulose |
| Quantity | 18 Tests/Kit |
| Substrate | Alkaline Phosphatase |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction