missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals HTLV Blot 2.4 (CE)

Product Code. p-200136117 Shop All MP Biomedicals Products
Click to view available options
Quantity:
18 Tests/Kit
36 Tests/Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30694933

Brand: MP Biomedicals 11080018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Blot is a qualitative enzyme immunoassay for the in vitro detection of antibodies to HTLV-1 and HTLV-2 in human serum or plasma.

  • The nitrocellulose strips are precoated with HTLV viral proteins derived from native inactivated viral particles and genetically engineered proteins.
  • Individual nitrocellulose strips are incubated with diluted serum or plasma specimens and controls. Specific antibodies to HTLV-I/II, if present in the specimen will bind to the HTLV-I/II proteins on the strips.
  • The strips are washed to remove unbound materials while antibodies that bind specifically to the HTLV proteins can be visualized using a series of reactions with goat anti-human IgG conjugated with alkaline phosphatase and the substrate, BCIP/NBT.
  • Each strip also includes an internal sample addition control to minimize the risk of false negatives due to operational errors.
  • The possible serological profiles defined by the test include the following: HTLV Seropositive, HTLV-I Seropositive, HTLV-II Seropositive, Seronegative or Indeterminate.
  • It is intended for use as a more specific supplemental test on human serum or plasma specimens found repeatedly reactive using screening procedures like ELISA.
  • CE-marked.
  • For professional use only.
  • Human serum and plasma.

Specifications

Product Type Western Blot Kit
Detection Method Colorimetric
For Use With (Application) Immunoblot (Western)
Membrane Compatibility Nitrocellulose
Quantity 18 Tests/Kit
Substrate Alkaline Phosphatase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.