missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTF9C Rabbit anti-Human, Clone: 2U3W8, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | HTF9C |
|---|---|
| Clone | 2U3W8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18378255
|
Novus Biologicals
NBP3-15244-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18387104
|
Novus Biologicals
NBP3-15244-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HTF9C Monoclonal antibody specifically detects HTF9C in Human samples. It is validated for Western BlotSpecifications
| HTF9C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 27037 | |
| IgG | |
| Affinity purified |
| 2U3W8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HTF9CHpaII tiny fragments locus 9c protein, MGC102728, TRM2 tRNA methyltransferase 2 homolog A (S. cerevisiae), tRNA (uracil-5-)-methyltransferase homolog A, tRNA methyltransferase 2 homolog A | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HTF9C (Q8IZ69). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title