missing translation for 'onlineSavingsMsg'
Learn More

HTF9C Antibody (4C10-1A11), Novus Biologicals™

Product Code. 18394859 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394859 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394859 Supplier Novus Biologicals Supplier No. H00027037M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HTF9C Monoclonal antibody specifically detects HTF9C in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTF9C
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4C10-1A11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH13352
Gene Alias HTF9CHpaII tiny fragments locus 9c protein, MGC102728, TRM2 tRNA methyltransferase 2 homolog A (S. cerevisiae), tRNA (uracil-5-)-methyltransferase homolog A, tRNA methyltransferase 2 homolog A
Host Species Mouse
Immunogen TRMT2A (AAH13352.2, 1 a.a. ~ 625 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSENLDNEGPKPMESCGQESSSALSCPTVSVPPAAPAALEEVEKEGAGAATGPGPQPGLYSYIRDDLFTSEIFKLELQNVPRHASFSDVRRFLGRFGLQPHKTKLFGQPPCAFVTFRSAAERDKALRVLHGALWKGRPLSVRLARPKADPMARRRRQEGESEPPVTRVADVVTPLWTVPYAEQLERKQLECEQVLQKLAKEIGSTNRALLPWLLEQRHKHNKACCPLEGVRPSPQQTEYRNKCEFLVGVGVDGEDNTVGCRLGKYKGGTCAVAAPFDTVHIPEATKQVVKAFQEFIRSTPYSAYDPETYTGHWKQLTVRTSRRHQAMAIAYFHPQKLSPEELAELKTSLAQHFTAGPGRASGVTCLYFVEEGQRKTPSQEGLPLEHVAGDRCIHEDLLGLTFRISPHAFFQVNTPAAEVLYTVIQDWAQLDAGSMVLDVCCGTGTIGLALARKVKRVIGVELCPEAVEDARVNAQDNELSNVEFHCGRAEDLVPTLVSRLASQHLVAILDPPRAGLHSKVILAIRRAKNLRRLLYVSCNPRAAMGNFVDLCRAPSNRVKGIPFRPVKAVAVDLFPQTPHCEMLILFERVEHPNGTGVLGPHSPPAQPTPGPPDNTLQETGTFPSS
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 27037
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.