missing translation for 'onlineSavingsMsg'
Learn More

HTBA, MedChemExpress

Product Code. 16451796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451796 500 mg 500mg
16436996 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451796 Supplier Medchem Express Supplier No. HY15919
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress HTBA(3-Hydroxy-2,4,6-triiodobenzoic acid) for your research needs.

HTBA(3-Hydroxy-2,4,6-triiodobenzoic acid) for your research needs.

Specifications

Chemical Name or Material HTBA
CAS 53279-72-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H3I3O3
Quantity 500 mg
Solubility Information DMSO : ≥ 33 mg/mL (63.98 mM)
SMILES O=C(O)C1=C(I)C=C(I)C(O)=C1I
Molecular Weight (g/mol) 515.81
Formula Weight 515.81
Percent Purity 99.61%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.