missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HT1080 (human fibrosarcoma) Whole cell lysate, Non-denatured; Abnova
Non-denatured whole cell lysate of HT1080 (human fibrosarcoma)
Brand: Abnova L051V2.200ug
Additional Details : Weight : 0.02000kg
Description
- Tissue: Connective Tissue, Fibrosarcoma
- Host: Human
- Lysis Buffer: Modified RIPA lysis buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
- Storage buffer: In modified RIPA lysis buffer
Western Blotting, Immunoprecipitation
Specifications
Immunoprecipitation, Western Blot | |
Whole cell lysate (non-denatured) | |
Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). | |
200 μg | |
Human |
Connective Tissue, Fibrosarcoma | |
Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
In modified RIPA Lysis Buffer. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |