Learn More
Trajan Scientific and Medical™ HT Science Silicon Septa
Product Code. 15208623
Change view
Click to view available options
Diameter (Metric) Inner:
9 mm
11.5 mm
12.5 mm
Unit Size:
Pack of 24
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
15208623 12.5 mm Pack of 24
15288613 9 mm Pack of 25
15298613 11.5 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15208623 Supplier Trajan Scientific and Medical™ Supplier No. 0418992
Only null left
Add to basket
Add to basket
Bleed and temperature optimized material.
  • Bleed and temperature optimized septa
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 400°C (330°C for 17mm size)
Specifications
Type HT
Diameter (Metric) Inner 12.5 mm
Quantity 24/Pk.