Learn More
Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup Reagent
Removes excess primers and dNTPs. Does not interfere with downstream applications. Ideal for high-throughput PCR product cleanup using liquid handling platforms and multi-channel pipettes.
Brand:  Applied Biosystems™ 785954PK
12280.61 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
 View more versions of this product
Product Code. 15503697
  • £18420.00 / 23040 reactions
Explore more special offers
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Description
  • Mixture of Exonuclease I and Shrimp Alkaline Phosphatase for the enzymatic removal of excess primers and dNTPs following a PCR reaction
  • One-tube PCR cleanup
  • Shortened 14-minute protocol
  • Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at −20°C. Stable for one month at 4°C
23,040 reactions, 48 × 8-tube strips in a tray