missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup

Product Code. 15869916
Click to view available options
No. of Reactions:
1000 reactions
5000 reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15869916

Brand: Applied Biosystems™ 785955000RXN

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

HT ExoSAP-IT Fast High-Throughput PCR Product Clean-up, with its shortened 14 minute protocol, is a highly stable, customized formulation of the original ExoSAP-IT reagent

  • 14 minute protocol is half the time of standard protocols
  • One-tube PCR clean-up - add HT ExoSAP-IT Fast reagent directly to PCR product
  • 100% sample recovery - no loss of PCR products regardless of the fragment size
  • Stable at 4°C for one month
  • Removes excess primers and dNTPs - does not interfere with downstream applications sequencing, SNP analysis, single base extension, fragment analysis
  • It is ideal for high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes
  • Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Format 8 Tube Strip, 96-well Plate, Vial
Final Product Type Fast High-Throughput PCR Product Clean-up
For Use With (Application) For high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes
No. of Reactions 5000 reactions
Product Type HT ExoSAP-IT™ Fast High-Throughput PCR Product Clean-up
Quantity Each
Test Time 14 min

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.