missing translation for 'onlineSavingsMsg'
Learn More

BTX™ HT 96-well Electroporation Plate
GREENER_CHOICE

Product Code. p-7250834
Click to view available options
Size (Metric) Gap:
2 mm
4 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15477310

Brand: BTX™ 450450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HT 96-well Electroporation Plate for use with ECM 630 and ECM 830 Electroporation Systems.

  • 96-well disposable electroporation plate
  • 2 mm gap
  • 100 μL
  • Coated
  • 1 plate

Specifications

Format 96-well
For Use With (Application) Transfection of Mammalian Cell, Transformation of Bacteria and Yeast, siRNA and cDNA Library Screening
For Use With (Equipment) ECM 630 HT, ECM 830 HT, Gemini X2 HT, 96-Well Electroporation Systems
Product Line BTX Electroporation
Product Type Electroporation Consumable
Description 96-well Disposable Electroporation Plate, 2 mm gap, 100 μl, Coated, 1 plate
Electroporation Chamber 96-well Electroporation Plate
Quantity 1 Ea.
Size (English) Gap 0.078 in.
Size (Metric) Gap 2 mm
Volume (Metric) 100 μL (Working)
Sterility Sterile
Connection Type Banana to Pin Tip
Pulse Form Exponential Decay Wave, Square Wave
High-throughput Compatibility Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.