missing translation for 'onlineSavingsMsg'
Learn More

HSU79274 Antibody (2G4-1A1), Novus Biologicals™

Product Code. 18320819 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320819 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320819 Supplier Novus Biologicals Supplier No. H00029902M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSU79274 Monoclonal antibody specifically detects HSU79274 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSU79274
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 2G4-1A1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH20967
Gene Alias chromosome 12 open reading frame 24, HSU79274, hypothetical protein LOC29902, protein predicted by clone 23733
Host Species Mouse
Immunogen C12orf24 (AAH20967, 1 a.a. ~ 273 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLGQLLPHTARGLGAAEMPGQGPGSDWTERSSSAEPPAVAGTEGGGGGSAGYSCYQNSKGSDRIKDGYKVNSHIAKLQELWKTPQNQTIHLSKSMMEASFFKHPDLTTGQKRYLCSIAKIYNANYLKMLMKRQYMHVLQHSSQKPGVLTHHRSRLSSRYSQKQHYPCTTWRHQLEREDSGSSDIAAASAPEMLIQHSLWRPVRNKEGIKTGYASKTRCKSLKIFRRPRKLFMQTVSSDDSESHMSEEKKEEDLLNNFMQSMSIEEQGEHLMLT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 29902
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.