Learn More
Waters Corporation HSS T3 Method Transfer Kit
Product Code. 17112992
Encompass
Click to view available options
Particle Size:
1.8 to 3.5 μm
1.8 to 5 μm
Type:
Method Transfer Kit
Method Transfer Kit, High Res
Unit Size:
Pack of 2
Product Code. 17112992
Brand: Waters Corporation 186004976
Method transfer kits are designed to preserve the integrity of a separation as it is transferred between UPLC and HPLC platforms.
  • Based on the concept of maintaining column length [L] to particle size [dp] ratio [L/dp], these kits provide an ACQUITY UPLC column with an HPLC column of equivalent selectivity and resolving power.
  • Using the ACQUITY UPLC columns calculator, methods can be fully transferred from HPLC to UPLC or from UPLC to HPLC.
Specifications
Particle Shape Spherical
Particle Size 1.8 to 3.5 μm
Pore Size 100 Å
Quantity 2/Pk.
USP Type L1
Endcapped Yes
Type Method Transfer Kit, High Res