missing translation for 'onlineSavingsMsg'
Learn More

HSR6071, MedChemExpress

Product Code. p-200100649
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16482388

Brand: Medchem Express HYU00133

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HSR6071, a pyrazinecarboxamide derivative, is an orally active and potent antiallergic agent. HSR6071 potently inhibits the experimental asthma in rat models.

HSR6071, a pyrazinecarboxamide derivative, is an orally active and potent antiallergic agent. HSR6071 potently inhibits the experimental asthma in rat models.

Specifications

Chemical Name or Material HSR6071
CAS 111374-21-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C10H12N8O
Quantity 5 mg
Solubility Information H2O : 50 mg/mL (192.12 mM; ultrasonic and adjust pH to 12 with NaOH)
SMILES O=C(C1=NC(N2CCCC2)=CN=C1)NC3=NN=NN3
Molecular Weight (g/mol) 260.26
Formula Weight 260.26
Percent Purity 98.51%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.