missing translation for 'onlineSavingsMsg'
Learn More

HSR6071, MedChemExpress

Product Code. 16482388
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482388 5 mg 5mg
16480189 10 mg 10mg
16470706 20 mg 20mg
16423179 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482388 Supplier Medchem Express Supplier No. HYU00133
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress HSR6071, a pyrazinecarboxamide derivative, is an orally active and potent antiallergic agent. HSR6071 potently inhibits the experimental asthma in rat models.

HSR6071, a pyrazinecarboxamide derivative, is an orally active and potent antiallergic agent. HSR6071 potently inhibits the experimental asthma in rat models.

Specifications

Chemical Name or Material HSR6071
CAS 111374-21-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C10H12N8O
Quantity 5 mg
Solubility Information H2O : 50 mg/mL (192.12 mM; ultrasonic and adjust pH to 12 with NaOH)
SMILES O=C(C1=NC(N2CCCC2)=CN=C1)NC3=NN=NN3
Molecular Weight (g/mol) 260.26
Formula Weight 260.26
Percent Purity 98.51%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.