missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPH1/HSP105 Antibody (J1G12), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04304AF488
This item is not returnable.
View return policy
Description
HSPH1/HSP105 Monoclonal specifically detects HSPH1/HSP105 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| HSPH1/HSP105 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HSPH1 | |
| Recombinant human HSP105 alpha (1-858 aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Anti-human HSP105a mAb, clone J1G12, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human HSP105a protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA | |
| J1G12 | |
| Western Blot, ELISA | |
| Antigen NY-CO-25, heat shock 105kD beta, heat shock 105kDa protein 1, heat shock 105kDa/110kDa protein 1, Heat shock 110 kDa protein, heat shock protein 105 kDa, HSP105A, HSP105B, HSP105heat shock 105kD alpha, HSP110, KIAA0201DKFZp686M05240, NY-CO-25 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 10808 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction