missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation HSPgel HT 6 Column
Click to view available options
Solvents:
ODCB
THF
Unit Size:
Each
Description
- They offer fast, accurate and precise molecular weight polymer determinations as well as increased sample throughput and greatly reduced solvent consumption and disposal costs.
- With the advent of today's higher performance solvent management systems, many polymers can now be analyzed accurately and reproducibly utilizing lower volume columns.
- This results in total analysis times of 7 minutes or less, thereby increasing sample throughput up to 6x while decreasing solvent usage from 35-50 mL to 5 mL or less per run.
Specifications
Specifications
| Diameter (Metric) | 6 mm |
| Length (Metric) | 150 mm |
| Particle Shape | Spherical |
| Particle Size | 5 μm |
| Quantity | 1/Ea. |
| Solvents | THF |
| USP Type | L21 |
| Type | Column |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction