missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPC132 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98141AF488
This item is not returnable.
View return policy
Description
HSPC132 Polyclonal specifically detects HSPC132 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HSPC132 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| 15E1.1, MDM35, MDM-35, Mitochondrial Distribution And Morphology 35 Homolog, P53CSV, P53-Inducible Cell-Survival Factor, Protein 15E1.1, TP53 regulated inhibitor of apoptosis 1, TP53-Regulated Inhibitor Of Apoptosis, TRIAP1, WF-1 | |
| Produced in rabbits immunized with E. coli-derived Human HSPC132 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 51499 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction