missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPBAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530817
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (95%), Rat (96%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HSPBAP1 (HSPB (heat shock 27kDa) associated protein 1), also known as PASS1 (protein associated with small stress protein 1), is a 488 amino acid cytoplasmic protein that contains one JMJC (Jumonji C) domain and shares 80% identity with its rat homolog, PASS1. Widely expressed with highest expression in ovary, thymus and pancreas, HSPBAP1 is thought to play a role in mediating cellular stress responses within the cell. Due to the presence of a JMJC domain, HSPBAP1 may be involved in chromatin remodeling events. Defects or translocations in the gene encoding HSPBAP1 are associated with renal cell carcinoma 1 (RCC1), suggesting a possible role for HSPBAP1 in carcinogenesis. Three isoforms of HSPBAP1 exist due to alternative splicing events.
Specifications
| HSPBAP1 | |
| Polyclonal | |
| Unconjugated | |
| HSPBAP1 | |
| 27 kDa heat shock protein-associated protein 1; 3830421G21Rik; FLJ22623; FLJ39386; Heat shock protein; HSP; HSP27; HSP28; HSPB (heat shock 27kDa) associated protein 1; Hspb associated protein 1; HSPB1 associated protein 1; HSPB1-associated protein 1; HSPBAP1; PASS1; protein associated with small stress protein 1; protein associating with small stress protein PASS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96EW2 | |
| HSPBAP1 | |
| Recombinant protein encompassing a sequence within the center region of human HSPBAP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction